2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol ≥98%

Supplier: Matrix Scientific

007341-1G 007341-5G
101792-496EA 93.85 USD
101792-496 101792-498
2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol ≥98%
2-Quinolin-2-yl-1-(3-trifluoromethylphenyl)ethanol

Hazard: Irritant

Formula: C₁₈H₁₄F₃NO
Melting Pt: 103…105 °C
MDL Number: MFCD03424492
CAS Number: 502625-52-7

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR