Pyridazine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Pyridazinecarboxylic acid
007329-1G
007329-5G
101792-466EA
69.23
USD
101792-466
101792-468
Pyridazine-4-carboxylic acid ≥97%
Pyridazine-4-carboxylic acid
Hazard: Irritant
Formula:
C₅H₄N₂O₂ MW: 124.10 g/mol Melting Pt: 237-240°C |
MDL Number:
MFCD00011576 CAS Number: 50681-25-9 |
Specification Test Results
Melting point | 237-240°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...