1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol

Supplier: Matrix Scientific

007302-1G 007302-5G
101792-400EA 113.85 USD
101792-400 101792-402
1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol
1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol

Hazard: Irritant

Formula: C₁₈H₁₇NO₂
MW: 279.34 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD03452779
CAS Number: 94004-77-0

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR