1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol

Supplier: Matrix Scientific

007301-1G 007301-5G
101792-396EA 109.23 USD
101792-396 101792-398
1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol
1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂
Melting Pt: 109…111 °C
MDL Number: MFCD03426057
CAS Number: 185848-10-6

Order Now

Specification Test Results

Melting point 109-111°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR