1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol

Supplier: Matrix Scientific

007300-1G 007300-5G
101792-392EA 100 USD
101792-392 101792-394
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₂
Melting Pt: 132…134 °C
MDL Number: MFCD03426056
CAS Number: 849021-36-9

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR