2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Quinolin-2-yl)-1-(4-(trifluoromethyl)phenyl)ethanol

007299-1G 007299-5G
101792-388EA 113.85 USD
101792-388 101792-390
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol ≥97%
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol

Hazard: Irritant

Formula: C₁₈H₁₄F₃NO
Melting Pt: 171…173 °C
MDL Number: MFCD03424493
CAS Number: 496947-30-9

Order Now

Specification Test Results

Melting point 171-173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR