2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol ≥98%

Supplier: Matrix Scientific

007296-1G 007296-5G
101792-376EA 176.92 USD
101792-376 101792-378
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol ≥98%
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol

Hazard: Irritant

Formula: C₁₈H₁₄F₃NO
MW: 317.31 g/mol
Melting Pt: 141…143 °C
MDL Number: MFCD03424494
CAS Number: 493024-38-7

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR