2-Benzoxazol-2-yl-1-(4-trifluoromethylphenyl)ethanol ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(1,3-Benzoxazol-2-yl)-1-[4-(trifluoromethyl)phenyl]ethan-1-ol

007295-1G 007295-5G
101792-372EA 83.08 USD
101792-372 101792-374
2-Benzoxazol-2-yl-1-(4-trifluoromethylphenyl)ethanol ≥98%
2-Benzoxazol-2-yl-1-(4-trifluoromethylphenyl)ethanol

Hazard: Irritant

Formula: C₁₆H₁₂F₃NO₂
MW: 307.27 g/mol
Melting Pt: 193…196 °C
MDL Number: MFCD03424486
CAS Number: 502625-50-5

Order Now

Specification Test Results

Melting point 193-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR