2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol
Supplier: Matrix Scientific
007294-1G
007294-5G
101792-368EA
100
USD
101792-368
101792-370
2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol
2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol
Hazard: Irritant
Formula:
C₁₉H₂₁NO₂ Melting Pt: 147…149 °C |
MDL Number:
MFCD03426055 CAS Number: 849021-34-7 |
Specification Test Results
Melting point | 147-149°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...