2-Pyrazin-2-yl-1-(4-trifluoromethylphenyl)ethanol

Supplier: Matrix Scientific

007293-1G 007293-5G
101792-364EA 72.31 USD
101792-364 101792-366
2-Pyrazin-2-yl-1-(4-trifluoromethylphenyl)ethanol
2-Pyrazin-2-yl-1-(4-trifluoromethylphenyl)ethanol

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O
Melting Pt: 100…102 °C
MDL Number: MFCD03424491
CAS Number: 502625-51-6

Order Now

Specification Test Results

Melting point 100-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR