1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol

Supplier: Matrix Scientific

007292-1G 007292-5G
101792-360EA 104.62 USD
101792-360 101792-362
1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol
1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol

Hazard: Irritant

Formula: C₁₆H₂₀N₂O
Melting Pt: 97…99 °C
MDL Number: MFCD03426054
CAS Number: 849021-31-4

Order Now

Specification Test Results

Melting point 97-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR