1-Phenyl-2-(quinoxalin-2-yl)ethanol

Supplier: Matrix Scientific

007290-1G 007290-5G
101792-352EA 150.77 USD
101792-352 101792-354
1-Phenyl-2-(quinoxalin-2-yl)ethanol
1-Phenyl-2-(quinoxalin-2-yl)ethanol

Hazard: Irritant

Formula: C₁₆H₁₄N₂O
MW: 250.3 g/mol
Melting Pt: 84…87 °C
MDL Number: MFCD03426052
CAS Number: 849021-27-8

Order Now

Specification Test Results

Melting point 84-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR