1-Phenyl-2-(pyrazin-2-yl)ethanol

Supplier: Matrix Scientific

007289-1G 007289-5G
101792-348EA 95.38 USD
101792-348 101792-350
1-Phenyl-2-(pyrazin-2-yl)ethanol
1-Phenyl-2-(pyrazin-2-yl)ethanol

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD00235147
CAS Number: 36914-69-9

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR