2-(3,4-Dichlorophenyl)malondialdehyde

Supplier: Matrix Scientific

007288-1G 007288-5G
101792-344EA 100 USD
101792-344 101792-346
2-(3,4-Dichlorophenyl)malondialdehyde
2-(3,4-Dichlorophenyl)malondialdehyde

Hazard: Irritant

Formula: C₉H₆Cl₂O₂
Melting Pt: 167…170 °C
MDL Number: MFCD02261891
CAS Number: 849936-29-4

Order Now

Specification Test Results

Melting point 167-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR