2-(3-Methylphenyl)malondialdehyde

Supplier: Matrix Scientific

007284-1G 007284-5G
101792-332EA 98.46 USD
101792-332 101792-334
2-(3-Methylphenyl)malondialdehyde
2-(3-Methylphenyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₂
Melting Pt: 81…83 °C
MDL Number: MFCD03426050
CAS Number: 849021-24-5

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR