Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate

Supplier: Matrix Scientific

007275-1G 007275-5G
101792-312EA 49.23 USD
101792-312 101792-314
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate
Methyl-4-(3-methoxy-3-oxopropanoyl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₂O₅
Melting Pt: 91 °C
MDL Number: MFCD00800637
CAS Number: 22027-52-7

Order Now

Specification Test Results

Melting point 83-86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR