4-Thiazolecarboxylic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
Thiazole-4-carboxylic acid
007268-1G
007268-5G
101792-294EA
15.38
USD
101792-294
101792-296
4-Thiazolecarboxylic acid ≥98%
4-Thiazolecarboxylic acid
Hazard: Irritant
Formula:
C₄H₃NO₂S MW: 129.1371 g/mol Melting Pt: 197-199°C |
MDL Number:
MFCD00623589 CAS Number: 3973-08-8 |
Specification Test Results
Melting point | 197-199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...