4,4-Difluoropiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4,4-Difluoropiperidine HCl

007247-1G 007247-5G 007247-25G
101792-244EA 53.85 USD
101792-244 101792-246 101792-248
4,4-Difluoropiperidine hydrochloride ≥95%
4,4-Difluoropiperidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClF₂N
MW: 157.59 g/mol
Melting Pt: ca 172°C
MDL Number: MFCD03095381
CAS Number: 144230-52-4

Order Now

Specification Test Results

Melting point ca 172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR