2-Aminoimidazole hemisulfate ≥98%

Supplier: Matrix Scientific

007232-5G 007232-25G
101792-214EA 41.54 USD
101792-214 101792-216
2-Aminoimidazole hemisulfate ≥98%
2-Aminoimidazole hemisulphate

Hazard: Irritant/Store in Freezer

Formula: C₆H₁₂N₆O₄S
Melting Pt: 252°C (dec)
MDL Number: MFCD00013162
CAS Number: 1450-93-7

Order Now

Specification Test Results

Melting point 252°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR