6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin

Supplier: Matrix Scientific

Synonyms: 6-Chloro-7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one

007211-1G 007211-5G
101792-200EA 52.31 USD
101792-200 101792-202
6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin
6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin

Hazard: Irritant

Formula: C₁₀H₄ClF₃O₃
MW: 264.59 g/mol
Melting Pt: 178…182 °C
MDL Number: MFCD00068095
CAS Number: 119179-66-7

Order Now

Specification Test Results

Melting point 178-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR