Methyl-3-(chlorosulfonyl)thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(chlorosulphonyl)thiophene-2-carboxylate, 2-Carbomethoxy-3-thiophenesulphonyl chloride

007148-5G 007148-25G
101792-084EA 15.38 USD
101792-084 101792-086
Methyl-3-(chlorosulfonyl)thiophene-2-carboxylate ≥95%
Methyl-3-(chlorosulfonyl)thiophene-2-carboxylate

TCA listed: T
Hazard: Irritant

Formula: C₆H₅ClO₄S₂
MW: 240.6845 g/mol
Melting Pt: 63…64 °C
MDL Number: MFCD00068160
CAS Number: 59337-92-7

Order Now

Specification Test Results

Melting point 63-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR