Tropylium tetrafluoroborate ≥95%

Supplier: Matrix Scientific

Synonyms: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

007097-5G 007097-25G
101792-012EA 103.08 USD
101792-012 101792-014
Tropylium tetrafluoroborate ≥95%
Tropylium tetrafluoroborate

TCA listed: T
Hazard: Irritant/Moisture Sensitive/Store in Freezer

Formula: C₇H₇BF₄
MW: 177.94 g/mol
Melting Pt: ca 240°C (dec)
MDL Number: MFCD00013402
CAS Number: 27081-10-3

Order Now

Specification Test Results

Melting point ca 240°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR