4-Hydroxy-L-isoleucine ≥97%

Supplier: Matrix Scientific

Synonyms: L-4-Hydroxyisoleucine, H-Ile(4-OH)-OH, (2S,3R)-2-Amino-4-hydroxy-3-methylpentanoic acid, 4-Hydroxyisoleucine

007079-25MG 007079-100MG
101791-976EA 86.15 USD
101791-976 101791-978
4-Hydroxy-L-isoleucine ≥97%
4-Hydroxy-L-isoleucine

Hazard: Irritant

Formula: C₆H₁₃NO₃
MW: 147.18 g/mol
Melting Pt: >200 °C
MDL Number: MFCD06799350
CAS Number: 781658-23-9

Order Now

Specification Test Results

Melting point >200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR