4,5-Dimethoxy-2-nitrobenzonitrile ≥98%

Supplier: Matrix Scientific

007073-5G 007073-25G
101791-964EA 33.85 USD
101791-964 101791-966
4,5-Dimethoxy-2-nitrobenzonitrile ≥98%
4,5-Dimethoxy-2-nitrobenzonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₈N₂O₄
MW: 208.170 g/mol
Melting Pt: 195-197°C
MDL Number: MFCD00017564
CAS Number: 102714-71-6

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR