4-Chloro-3-(trifluoromethyl)pyridine hydrochloride ≥97%

Supplier: Matrix Scientific

007071-1G 007071-5G
101791-960EA 253.85 USD
101791-960 101791-962
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride ≥97%
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant/Irritant-Harmful

Formula: C₆H₄Cl₂F₃N
Melting Pt: 124-126°C
MDL Number: MFCD04972770
CAS Number: 732306-24-0

Order Now

Specification Test Results

Melting point 124-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR