4-tert-Butoxyphenol ≥98%

Supplier: Matrix Scientific

Synonyms: Hydroquinone Mono-tert-butyl Ether

007064-1G 007064-5G 007064-25G
101791-938EA 60 USD
101791-938 101791-940 101791-942
4-tert-Butoxyphenol ≥98%
4-tert-Butoxyphenol

Hazard: Irritant

Formula: C₁₀H₁₄O₂
Melting Pt: 157…159 °C
MDL Number: MFCD02183556
CAS Number: 2460-87-9

Order Now

Specification Test Results

Melting point 157-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR