2,8-Bis(trifluoromethyl)quinolin-4-ol ≥99%

Supplier: Matrix Scientific

006997-1G 006997-5G
101791-780EA 23.08 USD
101791-780 101791-782
2,8-Bis(trifluoromethyl)quinolin-4-ol ≥99%
2,8-Bis(trifluoromethyl)quinolin-4-ol

Hazard: Irritant

Formula: C₁₁H₅F₆NO
Melting Pt: 127…133 °C
MDL Number: MFCD00075091
CAS Number: 35853-41-9

Order Now

Specification Test Results

Melting point 127-133°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR