3-(Methylsulfonyl)aniline hydrochloride

Supplier: Matrix Scientific

006995-1G 006995-5G
101791-776EA 80 USD
101791-776 101791-778
3-(Methylsulfonyl)aniline hydrochloride
3-(Methylsulphonyl)aniline hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO₂S
Melting Pt: 228-230°C (dec)
MDL Number: MFCD00216487
CAS Number: 80213-28-1

Order Now

Specification Test Results

Melting point 228-230°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR