2-Fluoro-5-(methylsulfonyl)aniline

Supplier: Matrix Scientific

006993-1G 006993-5G
101791-768EA 36.92 USD
101791-768 101791-770
2-Fluoro-5-(methylsulfonyl)aniline
2-Fluoro-5-(methylsulfonyl)aniline

Hazard: Irritant

Formula: C₇H₈FNO₂S
Melting Pt: 70…72 °C
MDL Number: MFCD01941314
CAS Number: 387358-51-2

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR