6-(Trifluoromethyl)anthranilic acid

Supplier: Matrix Scientific

006992-1G 006992-5G
101791-764EA 110.77 USD
101791-764 101791-766
6-(Trifluoromethyl)anthranilic acid
6-(Trifluoromethyl)anthranilic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 200°C (sub)
MDL Number: MFCD01569538
CAS Number: 314-46-5

Order Now

Specification Test Results

Melting point >200°C(sub)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR