1-Acetyl-5-amino-2,3-dihydro-(1H)-indole

Supplier: Matrix Scientific

006991-1G 006991-5G
101791-760EA 169.23 USD
101791-760 101791-762
1-Acetyl-5-amino-2,3-dihydro-(1H)-indole
1-Acetyl-5-amino-2,3-dihydro-(1H)-indole

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
Melting Pt: 180…182 °C
MDL Number: MFCD00087208
CAS Number: 4993-96-8

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR