2-Aminoisophthalic acid ≥97%

Supplier: Matrix Scientific

006990-1G 006990-5G
101791-756EA 26.15 USD
101791-756 101791-758
2-Aminoisophthalic acid ≥97%
2-Aminoisophthalic acid

Hazard: Irritant

Formula: C₈H₇NO₄
Melting Pt: 302…304 [>350] °C
MDL Number: MFCD00817808
CAS Number: 39622-79-2

Order Now

Specification Test Results

Melting point 302-304 [>


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR