Ethyl-3-oxo-3-(pyridin-4-yl)propanoate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-oxo-3-(pyridin-4-yl)propanoate, Ethyl isonicotinoylacetate, Ethyl 3-oxo-3-(4-pyridyl)propionate

006955-1G 006955-5G
101791-666EA 21.54 USD
101791-666 101791-668
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate ≥98%
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 56…59 °C
MDL Number: MFCD00094021
CAS Number: 26377-17-3

Order Now

Specification Test Results

Melting point 56-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR