Methyl nicotinoylacetate

Supplier: Matrix Scientific

Synonyms: Methyl 3-oxo-3-(pyridin-3-yl)propanoate

006953-1G 006953-5G
101791-658EA 61.54 USD
101791-658 101791-660
Methyl nicotinoylacetate
Methyl nicotinoylacetate

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.175 g/mol
Melting Pt: 39…42 °C
MDL Number: MFCD00216524
CAS Number: 54950-20-8

Order Now

Specification Test Results

Melting point 39-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR