Methyl-4-trifluoromethylbenzoylacetate
Supplier: Matrix Scientific
Synonyms:
Methyl 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionoate, 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionic acid methyll ester, Methyl 4-(Trifluoromethyl)benzoylacetate, 4-(Trifluoromethyl)benzoylacetic acid methyll ester, Methyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate
006950-1G
006950-5G
101791-650EA
16.92
USD
101791-650
101791-652
Methyl-4-trifluoromethylbenzoylacetate
Methyl-4-trifluoromethylbenzoylacetate
Hazard: Irritant
Formula:
C₁₁H₉F₃O₃ MW: 246.185 g/mol Melting Pt: 58…61 °C |
MDL Number:
MFCD00216521 CAS Number: 212755-76-5 |
Specification Test Results
Melting point | 58-61°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...