Ethyl-3,4,5-trimethoxybenzoylacetate
Supplier: Matrix Scientific
006949-1G
006949-5G
101791-646EA
52.31
USD
101791-646
101791-648
Ethyl-3,4,5-trimethoxybenzoylacetate
Ethyl-3,4,5-trimethoxybenzoylacetate
Hazard: Irritant
Formula:
C₁₄H₁₈O₆ MW: 282.292 g/mol Melting Pt: 94…95 °C |
MDL Number:
MFCD00026908 CAS Number: 3044-56-2 |
Specification Test Results
Melting point | 94-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...