2-(4-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde

Supplier: Matrix Scientific

Synonyms: 4-(1,3-Dioxoprop-2-yl)-3-nitrobenzoic acid

006945-1G 006945-5G
101791-630EA 76.92 USD
101791-630 101791-632
2-(4-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde
2-(4-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₇NO₆
MW: 237.17 g/mol
Melting Pt: 199…201 °C (dec.)
MDL Number: MFCD00216499
CAS Number: 205985-96-2

Order Now

Specification Test Results

Melting point 199-201°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR