2-(5-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde monohydrate

Supplier: Matrix Scientific

006944-1G 006944-5G
101791-626EA 53.85 USD
101791-626 101791-628
2-(5-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde monohydrate
2-(5-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde monohydrate

Hazard: Irritant

Formula: C₁₀H₉NO₇
Melting Pt: 214…216 °C (dec.)
MDL Number: MFCD00216507
CAS Number: 205680-84-8

Order Now

Specification Test Results

Melting point 214-216°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR