2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde

Supplier: Matrix Scientific

Synonyms: 2-(2-Carboxypyridin-6-yl)malondialdehyde

006941-1G 006941-5G
101791-614EA 70.77 USD
101791-614 101791-616
2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde
2-(2-Hydroxycarbonyl-6-pyridyl)malondialdehyde

Hazard: Irritant

Formula: C₉H₇NO₄
MW: 193.16 g/mol
Melting Pt: 280 °C (dec.)
MDL Number: MFCD00216531
CAS Number: 212755-80-1

Order Now

Specification Test Results

Melting point 280°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR