Pyrimidine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

006932-1G 006932-5G
101791-592EA 35.38 USD
101791-592 101791-594
Pyrimidine-5-carboxylic acid ≥95%
Pyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C5H4N2O2
MW: 124.10 g/mol
Melting Pt: 273…275 °C (dec.)
MDL Number: MFCD00856162
CAS Number: 4595-61-3

Order Now

Specification Test Results

Melting point 273-275°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR