3-(3-Trifluoromethylphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid

006931-5G 006931-25G
101791-588EA 81.54 USD
101791-588 101791-590
3-(3-Trifluoromethylphenyl)propionic acid ≥98%
3-(3-Trifluoromethylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Melting Pt: 29…31 °C
MDL Number: MFCD00090487
CAS Number: 585-50-2

Order Now

Specification Test Results

Melting point 29-31°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR