2-(Methylsulfonyl)aniline hydrochloride

Supplier: Matrix Scientific

006909-1G 006909-5G
101791-534EA 60 USD
101791-534 101791-536
2-(Methylsulfonyl)aniline hydrochloride
2-(Methylsulphonyl)aniline hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO₂S
Melting Pt: 139…141 °C
MDL Number: MFCD00216484
CAS Number: 205985-95-1

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR