2-Chloro-4-(methylsulfonyl)aniline
Supplier: Matrix Scientific
006907-1G
006907-5G
101791-524EA
146.15
USD
101791-524
101791-526
2-Chloro-4-(methylsulfonyl)aniline
2-Chloro-4-(methylsulfonyl)aniline
Hazard: Irritant
Formula:
C₇H₈ClNO₂S Melting Pt: 192…194 °C |
MDL Number:
MFCD00152349 CAS Number: 13244-35-4 |
Specification Test Results
Melting point | 192-194°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...