4-(Methylsulfonyl)phenylhydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (4-(Methylsulphonyl)phenyl)hydrazine hydrochloride

006906-1G 006906-5G
101791-520EA 15.38 USD
101791-520 101791-522
4-(Methylsulfonyl)phenylhydrazine hydrochloride ≥95%
4-(Methylsulfonyl)phenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂S
Melting Pt: 202 °C (dec.)
MDL Number: MFCD00216494
CAS Number: 17852-67-4

Order Now

Specification Test Results

Melting point 202°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR