4-Methylsulfonylbenzylamine hydrochloride
Supplier: Matrix Scientific
Synonyms:
(4-(Methylsulphonyl)phenyl)methanamine hydrochloride
006905-1G
006905-5G
101791-516EA
29.23
USD
101791-516
101791-518
4-Methylsulfonylbenzylamine hydrochloride
4-Methylsulfonylbenzylamine hydrochloride
CAS: 4393-16-2
MDL Number: MFCD00216491
Molecular Formula: C8H12ClNO2S
Molecular Weight: 221,71
Hazard: Irritant
Formula:
C₈H₁₁NO₂S.HCl MW: 221.70 g/mol Melting Pt: 272…274 °C |
MDL Number:
MFCD00216491 CAS Number: 98593-51-2 |
Specification Test Results
Melting point | 272-274°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...