2-Methylsulfonyl-1-(2-trifluoromethylphenyl)ethanone
Supplier: Matrix Scientific
Synonyms:
2-(Methylsulphonyl)-2'-(trifluoromethyl)acetophenone
006904-1G
006904-5G
101791-512EA
44.62
USD
101791-512
101791-514
2-Methylsulfonyl-1-(2-trifluoromethylphenyl)ethanone
2-Methylsulfonyl-1-(2-trifluoromethylphenyl)ethanone
Hazard: Irritant
Formula:
C₁₀H₉F₃O₃S MW: 266.24 g/mol Melting Pt: 61…63 °C |
MDL Number:
MFCD01941307 CAS Number: 387350-34-7 |
Specification Test Results
Melting point | 61-63°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...