2-Methylsulfonyl-1-(4-trifluoromethylphenyl)ethanone

Supplier: Matrix Scientific

Synonyms: 4-(Methylsulphonylacetyl)benzotrifluoride

006903-1G 006903-5G
101791-508EA 40 USD
101791-508 101791-510
2-Methylsulfonyl-1-(4-trifluoromethylphenyl)ethanone
2-Methylsulfonyl-1-(4-trifluoromethylphenyl)ethanone

Hazard: Irritant

Formula: C₁₀H₉F₃O₃S
MW: 266.24 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD01941309
CAS Number: 386715-52-2

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR