1-(6-Methylpyridin-2-yl)-2-methylsulfonylethanone

Supplier: Matrix Scientific

Synonyms: 2-Methyl-6-[(methylsulphonyl)acetyl]pyridine

006901-1G 006901-5G
101791-500EA 49.23 USD
101791-500 101791-502
1-(6-Methylpyridin-2-yl)-2-methylsulfonylethanone
1-(6-Methylpyridin-2-yl)-2-methylsulfonylethanone

Hazard: Irritant

Formula: C₉H₁₁NO₃S
MW: 213.26 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD01941311
CAS Number: 386715-51-1

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR