1-(3-Fluorophenyl)-2-methylsulfonylethanone

Supplier: Matrix Scientific

Synonyms: 3'-Fluoro-2-(methylsulphonyl)acetophenone

006900-1G 006900-5G
101791-496EA 44.62 USD
101791-496 101791-498
1-(3-Fluorophenyl)-2-methylsulfonylethanone
1-(3-Fluorophenyl)-2-methylsulfonylethanone

Hazard: Irritant

Formula: C₉H₉FO₃S
MW: 216.23 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD01941312
CAS Number: 82652-12-8

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR