1-(5-Bromopyridin-3-yl)-2-methylsulfonylethanone

Supplier: Matrix Scientific

006899-1G 006899-5G
101791-492EA 52.31 USD
101791-492 101791-494
1-(5-Bromopyridin-3-yl)-2-methylsulfonylethanone
1-(5-Bromopyridin-3-yl)-2-methylsulfonylethanone

Hazard: Irritant

Formula: C₈H₈BrNO₃S
Melting Pt: 158…161 °C
MDL Number: MFCD01941313
CAS Number: 386715-50-0

Order Now

Specification Test Results

Melting point 158-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR